Alfa Aesar™ (1,1-Dimethoxyethyl)benzene, 97%

Manufacturer: Alfa Aesar™ H5569606

Quantity 5g

 View more versions of this product

Catalog No. AAH5569606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4316-35-2
    Formula Weight 166.22
    MDL Number MFCD00075617
    Chemical Name or Material (1,1-Dimethoxyethyl)benzene

    Specifications

    Boiling Point 95° to 98°C (12mmHg)
    Formula Weight 166.22
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Density 1.008g/mL
    Color Colorless
    MDL Number MFCD00075617
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Refractive Index 1.4900
    Physical Form Liquid
    CAS 4316-35-2
    Quantity 5g
    Chemical Name or Material (1,1-Dimethoxyethyl)benzene
    Assay 97%

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit