Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dimethyl-2-phenylethyl Acetate 99.0+%, TCI America™

Click to view available options
Quantity:
25g
151-05-3
C12H16O2
192.258
MFCD00026196
FLUWAIIVLCVEKF-UHFFFAOYSA-N
Acetic Acid alpha,alpha-Dimethylphenethyl Ester, alpha,alpha-Dimethylphenethyl Acetate, Acetic Acid 1,1-Dimethyl-2-phenylethyl Ester
9024
(2-methyl-1-phenylpropan-2-yl) acetate
CC(=O)OC(C)(C)CC1=CC=CC=C1
151-05-3
C12H16O2
192.258
MFCD00026196
FLUWAIIVLCVEKF-UHFFFAOYSA-N
Acetic Acid alpha,alpha-Dimethylphenethyl Ester, alpha,alpha-Dimethylphenethyl Acetate, Acetic Acid 1,1-Dimethyl-2-phenylethyl Ester
9024
(2-methyl-1-phenylpropan-2-yl) acetate
CC(=O)OC(C)(C)CC1=CC=CC=C1
Color White
Quantity 25g
Formula Weight 192.26
Physical Form Crystal at 20°C
Percent Purity ≥99.0% (GC)
Chemical Name or Material 1,1-Dimethyl-2-phenylethyl Acetate

EINECSNumber : (3)-1045

RTECSNumber : SG8100000

TSCA : Yes