Alfa Aesar™ 1,1'-Dimethylferrocene, 97%

Manufacturer: Alfa Aesar™ A1217506

Quantity 5g

 View more versions of this product

Catalog No. AAA1217506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1291-47-0
    Molecular Formula C12H14Fe
    Formula Weight 214.09
    MDL Number MFCD00001425
    Synonym Bis(methylcyclopentadienyl)iron
    Chemical Name or Material 1,1'-Dimethylferrocene

    Specifications

    Formula Weight 214.09
    Molecular Formula C12H14Fe
    Synonym Bis(methylcyclopentadienyl)iron
    Flash Point 108°C (226°F)
    Melting Point 36° to 41°C
    MDL Number MFCD00001425
    CAS 1291-47-0
    Quantity 5g
    Chemical Name or Material 1,1'-Dimethylferrocene
    Assay 97%

    Description

    Applications
    It is used as a combustion control additive in fuels, antiknock agent in gasoline, and for heat stabilization in greases and plastics. It is used as a catalyst for synthesis of ammonia, polymerization.

    Solubility
    Slightly soluble in methanol. Insoluble in water.

    Notes
    Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit