Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,1'-Dimethylferrocene, 97%

Manufacturer:  Alfa Aesar™ A1217503

Quantity 1g

 View more versions of this product

Catalog No. AAA1217503


  Check Availability
Add to cart

Specifications

Description

Applications
It is used as a combustion control additive in fuels, antiknock agent in gasoline, and for heat stabilization in greases and plastics. It is used as a catalyst for synthesis of ammonia, polymerization.

Solubility
Slightly soluble in methanol. Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Description & Specifications

Chemical Identifiers

1291-47-0
214.09
Bis(methylcyclopentadienyl)iron
C12H14Fe
MFCD00001425
1,1'-Dimethylferrocene

Specifications

108°C (226°F)
1g
C12H14Fe
MFCD00001425
1,1'-Dimethylferrocene
36° to 41°C
1291-47-0
214.09
Bis(methylcyclopentadienyl)iron
97%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit