Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dimethylurea, 98+%, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
598-94-7
C3H8N2O
88.11
MFCD00007959
YBBLOADPFWKNGS-UHFFFAOYSA-N
n,n-dimethylurea,urea, n,n-dimethyl,asym-dimethylurea,urea, dimethyl,urea, 1,1-dimethyl,n,n-dimethylharnstoff german,n,n-dimethylharnstoff,dimethyl urea,1.1-dimethylurea,1,1-dimethyl-urea
11737
1,1-dimethylurea
CN(C)C(=O)N
598-94-7
C3H8N2O
88.11
MFCD00007959
YBBLOADPFWKNGS-UHFFFAOYSA-N
n,n-dimethylurea,urea, n,n-dimethyl,asym-dimethylurea,urea, dimethyl,urea, 1,1-dimethyl,n,n-dimethylharnstoff german,n,n-dimethylharnstoff,dimethyl urea,1.1-dimethylurea,1,1-dimethyl-urea
11737
1,1-dimethylurea
CN(C)C(=O)N
Infrared Spectrum Authentic
Quantity 5g
Melting Point 179°C to 183°C
Color White
Packaging Glass bottle
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula (CH3)2NC(O)NH2
Solubility Information Solubility in water: soluble
Formula Weight 88.11
Physical Form Crystalline Powder
Percent Purity 98+%
Chemical Name or Material 1, 1-Dimethylurea
Show More Show Less

EINECSNumber : 209-957-