Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Diphenylethylene, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
530-48-3
C14H12
180.25
ZMYIIHDQURVDRB-UHFFFAOYSA-N
1,1-diphenylethylene,ethene-1,1-diyldibenzene,1,1-diphenylethene,benzene, 1,1'-ethenylidenebis,1-phenylethenyl benzene,unii-bx0l5b6lll,as-diphenylethylene,.alpha.-phenylstyrene,1-phenylvinyl benzene,bx0l5b6lll
10740
1-phenylethenylbenzene
C=C(C1=CC=CC=C1)C2=CC=CC=C2
530-48-3
C14H12
180.25
ZMYIIHDQURVDRB-UHFFFAOYSA-N
1,1-diphenylethylene,ethene-1,1-diyldibenzene,1,1-diphenylethene,benzene, 1,1'-ethenylidenebis,1-phenylethenyl benzene,unii-bx0l5b6lll,as-diphenylethylene,.alpha.-phenylstyrene,1-phenylvinyl benzene,bx0l5b6lll
10740
1-phenylethenylbenzene
C=C(C1=CC=CC=C1)C2=CC=CC=C2
Quantity 100g
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.02
Refractive Index 1.6065 to 1.6085
Color Colorless
Density 1.0200g/mL
Flash Point >112°C
Boiling Point 270.0°C to 271.0°C
Melting Point 6.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (C6H5)2C=CH2
Beilstein 05,639
Merck Index 15,3356
Formula Weight 180.25
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1, 1-Diphenylethylene
Show More Show Less