Alfa Aesar™ 1,1-Diphenylhexane, 98%

Manufacturer: Alfa Aesar™ B2289606

Quantity 5g

 View more versions of this product

Catalog No. AAB2289606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1530-04-7
    Molecular Formula (C6H5)2CH(CH2)4CH3
    Formula Weight 238.38
    MDL Number MFCD00041694
    Chemical Name or Material 1,1-Diphenylhexane

    Specifications

    Formula Weight 238.38
    Molecular Formula (C6H5)2CH(CH2)4CH3
    MDL Number MFCD00041694
    CAS 1530-04-7
    Boiling Point 129° to 130°C (2mmHg)
    Quantity 5g
    Chemical Name or Material 1,1-Diphenylhexane
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit