Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,1-Diphenylpentane, 97%

Click to view available options
Quantity:
5g
25g
1726-12-1
C17H20
224.347
MFCD00041692
GSGNVNZDHCKRGI-UHFFFAOYSA-N
1,1-diphenylpentane,benzene, 1,1'-pentylidenebis,1-phenylpentyl benzene,diphenylpentane,phenylpentyl benzene,acmc-20ap7o,pentane-1,1-diyldibenzene,benzene,1,1'-pentylidenebis
137174
1-phenylpentylbenzene
CCCCC(C1=CC=CC=C1)C2=CC=CC=C2
1726-12-1
C17H20
224.347
MFCD00041692
GSGNVNZDHCKRGI-UHFFFAOYSA-N
1,1-diphenylpentane,benzene, 1,1'-pentylidenebis,1-phenylpentyl benzene,diphenylpentane,phenylpentyl benzene,acmc-20ap7o,pentane-1,1-diyldibenzene,benzene,1,1'-pentylidenebis
137174
1-phenylpentylbenzene
CCCCC(C1=CC=CC=C1)C2=CC=CC=C2
Boiling Point 138°C to 139°C (15mmHg)
Quantity 5g
Formula Weight 224.35
Percent Purity 97%
Chemical Name or Material 1,1-Diphenylpentane