Alfa Aesar™ 1,1'-Ferrocenedicarboxaldehyde, 97%

Manufacturer: Alfa Aesar™ 4429202

Quantity 0.5g

 View more versions of this product

Catalog No. AA4429202

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1271-48-3
    Molecular Formula C12H10FeO2
    Formula Weight 242.06
    MDL Number MFCD02093629
    Chemical Name or Material 1,1'-Ferrocenedicarboxaldehyde

    Specifications

    Formula Weight 242.06
    Quantity 0.5g
    Physical Form Powder
    Sensitivity Air Sensitive
    Melting Point 183° to 184°C
    Molecular Formula C12H10FeO2
    MDL Number MFCD02093629
    CAS 1271-48-3
    Chemical Name or Material 1,1'-Ferrocenedicarboxaldehyde
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit