Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Ferrocenedimethanol, 98%, Acros Organics

Click to view available options
Quantity:
5g
1291-48-1
C12H14FeO2
ferrocene derivatives,1,1'-bis hydroxymethyl ferrocen,1,1'-ferrocenedimethanol,1,1 inverted exclamation marka-ferrocenedimethanol
1291-48-1
C12H14FeO2
ferrocene derivatives,1,1'-bis hydroxymethyl ferrocen,1,1'-ferrocenedimethanol,1,1 inverted exclamation marka-ferrocenedimethanol
Quantity 5g
Melting Point 106.0°C to 109.0°C
Packaging Glass bottle
Assay Percent Range 98%
Solubility Information Solubility in water: insoluble.
Formula Weight 246.09
Percent Purity 98%
Chemical Name or Material 1,1′-Ferrocenedimethanol