Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1-Methyl-4-piperidinyl)piperazine, 98%, Thermo Scientific™

Catalog Number AC389560010
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
23995-88-2
C10H21N3
183.299
OHUMKYGINIODOY-UHFFFAOYSA-N
1-1-methyl-4-piperidinyl piperazine, 1-1-methylpiperidin-4-yl piperazine, 1-n-methylpiperidin-4-yl piperazine, 1-1-methyl-piperidin-4-yl-piperazine, piperazine, 1-1-methyl-4-piperidinyl, 1-1-methyl-4-piperidyl piperazine, 1-1-methyl-4-piperidino piperazine, zlchem 509, acmc-1cq7x, 1-methyl-4-piperidyl piperazine
566324
1-(1-methylpiperidin-4-yl)piperazine
CN1CCC(CC1)N2CCNCC2
23995-88-2
C10H21N3
183.299
OHUMKYGINIODOY-UHFFFAOYSA-N
1-1-methyl-4-piperidinyl piperazine, 1-1-methylpiperidin-4-yl piperazine, 1-n-methylpiperidin-4-yl piperazine, 1-1-methyl-piperidin-4-yl-piperazine, piperazine, 1-1-methyl-4-piperidinyl, 1-1-methyl-4-piperidyl piperazine, 1-1-methyl-4-piperidino piperazine, zlchem 509, acmc-1cq7x, 1-methyl-4-piperidyl piperazine
566324
1-(1-methylpiperidin-4-yl)piperazine
CN1CCC(CC1)N2CCNCC2

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 112.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Refractive Index 1.5070 to 1.5090
Quantity 1 g
Formula Weight 183.3
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-(1-Methyl-4-piperidinyl)piperazine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit