Alfa Aesar™ 1,1'-Oxybis(3,3,3-trifluoro-1-propanol) hemihydrate, 96%

Manufacturer: Alfa Aesar™ H5169003

Quantity 1g

 View more versions of this product

Catalog No. AAH5169003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 936107-87-8
    Molecular Formula C6H8F6O3·0.5H2O
    Linear Formula [CF3CH2CH(OH)]2O·0.5H2O
    Formula Weight 251.12 (242.12 Anhydrous)
    MDL Number MFCD00156074
    UN Number UN1988
    Synonym 3,3,3-Trifluoropropionaldehyde hemihydrate
    Chemical Name or Material 1,1'-Oxybis(3,3,3-trifluoro-1-propanol) hemihydrate

    Specifications

    Formula Weight 251.12 (242.12 Anhydrous)
    Linear Formula [CF3CH2CH(OH)]2O·0.5H2O
    MDL Number MFCD00156074
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C6H8F6O3·0.5H2O
    CAS 936107-87-8
    UN Number UN1988
    Synonym 3,3,3-Trifluoropropionaldehyde hemihydrate
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H318-H226-H302-H315-H335
    Toxic if inhaled.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Causes skin irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, FLAMMABLE, TOXIC, N.O.S.
    Quantity 1g
    Chemical Name or Material 1,1'-Oxybis(3,3,3-trifluoro-1-propanol) hemihydrate
    Assay 96%

    GHS H Statement
    H331-H318-H226-H302-H315-H335
    Toxic if inhaled.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit