Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(1-Propynyl)cyclohexanol, 98%

Click to view available options
Quantity:
10g
50g
697-37-0
C9H14O
138.21
MFCD00078311
UTISCLIDFORRJP-UHFFFAOYSA-N
1-1-propynyl cyclohexanol,1-prop-1-yn-1-yl cyclohexan-1-ol,1-prop-1-yn-1-yl cyclohexanol,1-1-propynyl cyclohexanol 10g
230341
1-prop-1-ynylcyclohexan-1-ol
CC#CC1(CCCCC1)O
697-37-0
C9H14O
138.21
MFCD00078311
UTISCLIDFORRJP-UHFFFAOYSA-N
1-1-propynyl cyclohexanol,1-prop-1-yn-1-yl cyclohexan-1-ol,1-prop-1-yn-1-yl cyclohexanol,1-1-propynyl cyclohexanol 10g
230341
1-prop-1-ynylcyclohexan-1-ol
CC#CC1(CCCCC1)O
Melting Point 46°C to 50°C
Boiling Point 91°C to 92°C (10mmHg)
Quantity 10g
Beilstein 1856828
Formula Weight 138.21
Percent Purity 98%
Chemical Name or Material 1-(1-Propynyl)cyclohexanol