1,1'-Thiocarbonyldiimidazole, tech 90%

Manufacturer: Alfa Aesar B2200809

Quantity 10g

 View more versions of this product

Catalog No. AAB2200809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6160-65-2
    Molecular Formula C7H6N4S
    Formula Weight 178.22
    MDL Number MFCD00005289
    Beilstein 609349
    Chemical Name or Material 1, 1'-Thiocarbonyldiimidazole
    Grade Technical Grade

    Specifications

    Formula Weight 178.22
    Quantity 10g
    MDL Number MFCD00005289
    Beilstein 609349
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for se
    Recommended Storage 2° to 8°C
    Assay Percent Range 0.95
    Molecular Formula C7H6N4S
    CAS 6160-65-2
    Storage Note 1 Moisture Sensitive
    Chemical Name or Material 1, 1'-Thiocarbonyldiimidazole
    Assay 90%
    Grade Technical Grade

    Description

    Applications
    1,1 ft.-Thiocarbonyldiimidazole is used as a reagent for the deoxygenation of alcohols with tributyltin hydride and diols with lithium aluminum hydride. It is also used to deoxygenate carboxylic monosaccharide analogues. Further, it is a useful reagent involved in biochemical synthesis. In addition to this, it is used to prepare 1-[bis-(4-methoxy-phenyl)-methyl]-1H-imidazole by reacting with 4,4 ft.-dimethoxy-benzhydrol.

    Solubility
    Soluble in terahydrofuran, toluene and dichloromethane.

    Notes
    Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Store in a cool place. Incompatible with strong oxidizing agents and strong acids.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for se

    Moisture Sensitive

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit