Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Decanediol, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
112-47-0
C10H22O2
174.28
MFCD00004749
FOTKYAAJKYLFFN-UHFFFAOYSA-N
1,10-decanediol,decamethylene glycol,decamethylenediol,1,10-dihydroxydecane,1,10-decamethylenediol,1,10-decamethylene diol,1,6-bis 2-hydroxyethyl hexane,alpha,omega-decanediol,unii-5i577udk52,1,10-ddecanediol
37153
decane-1,10-diol
C(CCCCCO)CCCCO
112-47-0
C10H22O2
174.28
MFCD00004749
FOTKYAAJKYLFFN-UHFFFAOYSA-N
1,10-decanediol,decamethylene glycol,decamethylenediol,1,10-dihydroxydecane,1,10-decamethylenediol,1,10-decamethylene diol,1,6-bis 2-hydroxyethyl hexane,alpha,omega-decanediol,unii-5i577udk52,1,10-ddecanediol
37153
decane-1,10-diol
C(CCCCCO)CCCCO
Boiling Point 297°C
Flash Point 152°C
Melting Point 71°C to 75°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100g
Color White
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula HO(CH2)10OH
Beilstein 01, II, 560
Merck Index 15, 2853
Solubility Information Solubility in water: insoluble in water. Other solubilities: freely soluble in alcohol and warm ether,practically insoluble in petroleum ether
Formula Weight 174.28
Physical Form Pellets or Flakes and/or Powder
Percent Purity 99%
Chemical Name or Material 1, 10-Decanediol
Show More Show Less

EINECSNumber : 203-975-2