Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Diamino-4,7-dioxadecane, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass Bottle
2997-01-5
C8H22N2O2
178.28
MFCD00059938
POTQBGGWSWSMCX-UHFFFAOYSA-P
di 3-aminopropoxy ethane,1-propanamine, 3,3'-1,2-ethanediylbis oxy bis,1,2-bis 3-aminopropoxy ethane,3,3'-ethane-1,2-diylbis oxy bis propan-1-amine,3,3'-ethylenedioxy bispropylamine,3,3'-ethylenedioxybis propylamine,propylamine, 3,3'-ethylenedioxy bis,ethylene glycol bis 3-aminopropyl ether,1,10-diamino-4,7-dioxadecane,3-2-3-aminopropoxy ethoxy propan-1-amine
76342
3-[2-(3-aminopropoxy)ethoxy]propan-1-amine
C(CN)COCCOCCCN
2997-01-5
C8H22N2O2
178.28
MFCD00059938
POTQBGGWSWSMCX-UHFFFAOYSA-P
di 3-aminopropoxy ethane,1-propanamine, 3,3'-1,2-ethanediylbis oxy bis,1,2-bis 3-aminopropoxy ethane,3,3'-ethane-1,2-diylbis oxy bis propan-1-amine,3,3'-ethylenedioxy bispropylamine,3,3'-ethylenedioxybis propylamine,propylamine, 3,3'-ethylenedioxy bis,ethylene glycol bis 3-aminopropyl ether,1,10-diamino-4,7-dioxadecane,3-2-3-aminopropoxy ethoxy propan-1-amine
76342
3-[2-(3-aminopropoxy)ethoxy]propan-1-amine
C(CN)COCCOCCCN
Quantity 25g
Refractive Index 1.4620 to 1.4640 (20°C, 589nm)
Packaging Glass Bottle
Density 0.9837g/mL
Color Colorless
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula NH2(CH2)3O(CH2)2O(CH2)3NH2
Formula Weight 176.26
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material 1, 10-Diamino-4, 7-dioxadecane
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger