Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Diaza-18-crown-6, 96%, Thermo Scientific Chemicals

Catalog Number p-7033874
Click to view available options
Quantity:
250 mg
1 g
5 g
23978-55-4
C12H28N2O4
264.37
MFCD00005112
NLMDJJTUQPXZFG-UHFFFAOYSA-P
kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane
72805
1,4,10,13-tetraoxa-7,16-diazacyclooctadecane
C1COCCOCC[NH2+]CCOCCOCC[NH2+]1
23978-55-4
C12H28N2O4
264.37
MFCD00005112
NLMDJJTUQPXZFG-UHFFFAOYSA-P
kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane
72805
1,4,10,13-tetraoxa-7,16-diazacyclooctadecane
C1COCCOCC[NH2+]CCOCCOCC[NH2+]1

1,10-Diaza-18-crown-6 are starting materials to produce macrocyclic polyamine metal-complexing agents. It is used as a ligand in chemistry for the applications of diagnostic imaging technique to identify medical condition or disease and for the development of medication as a therapeutic agent and pharmaceutic aid. Also used as an ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 609764
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 262.35
Percent Purity 96%
Chemical Name or Material 1,10-Diaza-18-crown-6
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 245-965-0
RTECSNumber HM1650000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit