1,10-Dibromodecane, 97%, Alfa Aesar™

Manufacturer: Alfa Aesar A1235722

Quantity 100g

 View more versions of this product

Catalog No. AAA1235722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4101-68-2
    Molecular Formula Br(CH2)10Br
    Formula Weight 300.08
    MDL Number MFCD00000222
    Beilstein 506156
    Synonym Decamethylene dibromide
    Chemical Name or Material 1,10-Dibromodecane

    Specifications

    Formula Weight 300.08
    Melting Point 26° to 30°C
    Quantity 100g
    TSCA TSCA
    Molecular Formula Br(CH2)10Br
    MDL Number MFCD00000222
    CAS 4101-68-2
    Beilstein 506156
    Synonym Decamethylene dibromide
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335;
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a;
    Avoid breathing dust/fume/gas/mist/vapours/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 157° to 159°C (14mmHg)
    Flash Point >110°C (230°F)
    Chemical Name or Material 1,10-Dibromodecane
    Assay 97%

    Description

    Applications
    1,10-Dibromodecane is used as an alkylating agent and as synthetic organic intermediate.

    Solubility
    Insoluble in water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.

    GHS H Statement
    H315-H319-H335;
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a;
    Avoid breathing dust/fume/gas/mist/vapours/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit