Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Phenanthroline monohydrate, 99+%, pure, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
5144-89-8,66-71-7
C12H8N2·H2O
198.23
MFCD00149973
PPQJCISYYXZCAE-UHFFFAOYSA-N
1,10-phenanthroline hydrate,1,10-phenanthroline monohydrate,o-phenanthroline monohydrate,unii-ksx215x00e,1,10-fenantrolina,4,5-phenanthroline monohydrate,1,10-phenanthroline, monohydrate,1,10-fenanthrolin,1,10-phenanthrolineo-phenanthroline,1, 10-phenanthroline monohydrate
21226
1,10-phenanthroline;hydrate
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O
5144-89-8,66-71-7
C12H8N2·H2O
198.23
MFCD00149973
PPQJCISYYXZCAE-UHFFFAOYSA-N
1,10-phenanthroline hydrate,1,10-phenanthroline monohydrate,o-phenanthroline monohydrate,unii-ksx215x00e,1,10-fenantrolina,4,5-phenanthroline monohydrate,1,10-phenanthroline, monohydrate,1,10-fenanthrolin,1,10-phenanthrolineo-phenanthroline,1, 10-phenanthroline monohydrate
21226
1,10-phenanthroline;hydrate
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O
Indicator
Packaging Glass bottle
Quantity 5g
Color Cream-White
Infrared Spectrum Authentic
Melting Point 97.0°C to 101.0°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Beilstein 23,227
Merck Index 15,7328
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, acetone and benzene
Formula Weight 198.23
Physical Form Needle-Like Crystalline Powder
Percent Purity 99+%
Grade Pure
Chemical Name or Material 1, 10-Phenanthroline monohydrate
Show More Show Less

GHS H Statement
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

GHS Signal Word: Danger