Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Phenanthroline, Monohydrate, ACS, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5144-89-8
5144-89-8
Packaging Amber Glass Bottle
Quantity 1 kg
CAS Max % 1
Grade ACS

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.