Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Phenanthroline Monohydrate (Certified ACS), Fisher Chemical

Click to view available options
Quantity:
5g
10g
Packaging:
Glass Bottle
5144-89-8
C12H10N2O
198.225
MFCD00149973
PPQJCISYYXZCAE-UHFFFAOYSA-N
1,10-phenanthroline hydrate,1,10-phenanthroline monohydrate,o-phenanthroline monohydrate,unii-ksx215x00e,1,10-fenantrolina,4,5-phenanthroline monohydrate,1,10-phenanthroline, monohydrate,1,10-fenanthrolin,1,10-phenanthrolineo-phenanthroline,1, 10-phenanthroline monohydrate
21226
1,10-phenanthroline;hydrate
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O
5144-89-8
C12H10N2O
198.225
MFCD00149973
PPQJCISYYXZCAE-UHFFFAOYSA-N
1,10-phenanthroline hydrate,1,10-phenanthroline monohydrate,o-phenanthroline monohydrate,unii-ksx215x00e,1,10-fenantrolina,4,5-phenanthroline monohydrate,1,10-phenanthroline, monohydrate,1,10-fenanthrolin,1,10-phenanthrolineo-phenanthroline,1, 10-phenanthroline monohydrate
21226
1,10-phenanthroline;hydrate
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O
Packaging Glass Bottle
Alpha Vector Phenanthroline Monohydrate
Boiling Point 90.2°C (25mmHg)
Color White
Suitability Pass Test (as redox indicator and for Fe test)
Quantity 5g
Flash Point 79°C
Melting Point 97°C
UN Number 2811
Formula Weight 198.23
Physical Form Powder/Solid
Grade Certified ACS
Chemical Name or Material 1, 10-Phenanthroline Monohydrate
Show More Show Less

DOTInformation : DOT Class 6.1, : Poison