Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Phenanthroline Monohydrochloride Monohydrate, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
18851-33-7
HCl·H2O
234.69
MFCD00150061
NDLHUHRGAIHALB-UHFFFAOYSA-N
1,10-phenanthroline hydrochloride monohydrate,1,10-phenanthroline hydrochloride hydrate,1,10-phenanthroline monohydrochloride monohydrate,1,10-phenanthroline hydrate hydrochloride,1,10-phenanthroline, monohydrochloride, monohydrate,phen hydrate hydrochloride,acmc-209iz9,dsstox_cid_24309,dsstox_rid_80145,dsstox_gsid_44309
2723715
1,10-phenanthroline;hydrate;hydrochloride
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O.Cl
18851-33-7
HCl·H2O
234.69
MFCD00150061
NDLHUHRGAIHALB-UHFFFAOYSA-N
1,10-phenanthroline hydrochloride monohydrate,1,10-phenanthroline hydrochloride hydrate,1,10-phenanthroline monohydrochloride monohydrate,1,10-phenanthroline hydrate hydrochloride,1,10-phenanthroline, monohydrochloride, monohydrate,phen hydrate hydrochloride,acmc-209iz9,dsstox_cid_24309,dsstox_rid_80145,dsstox_gsid_44309
2723715
1,10-phenanthroline;hydrate;hydrochloride
C1=CC2=C(C3=C(C=CC=N3)C=C2)N=C1.O.Cl
Quantity 25g
Suitability suitable for determination of iron
Water 8.5% max. (K.F.)
Packaging Glass bottle
Sulfated Ash 0.1% max.
Infrared Spectrum Authentic
Color Purple or White to Yellow
Melting Point 224.0°C to 225.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Solubility Information Solubility in water: freely soluble.
Formula Weight 234.69
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 1, 10-Phenanthroline monohydrochloride monohydrate
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger