Alfa Aesar™ (1,10-Phenanthroline)(trifluoromethyl)copper(I), 95%

Manufacturer: Alfa Aesar™ 4706806

Quantity 5g

 View more versions of this product

Catalog No. AA4706806

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1300746-79-5
    Linear Formula (C12H8N2)CuCF3
    Formula Weight 312.76
    MDL Number MFCD19690901
    Synonym Trifluoromethyl(1,10-phenanthroline)copper(I); Trifluoromethylator|r
    Chemical Name or Material (1,10-Phenanthroline)(trifluoromethyl)copper(I)

    Specifications

    Formula Weight 312.76
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P301+P312-P312-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 1300746-79-5
    Color Orange
    Sensitivity Air & Moisture Sensitive
    MDL Number MFCD19690901
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H302-H312-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Storage Note 1 2° to 8°C
    Physical Form Solid
    Linear Formula (C12H8N2)CuCF3
    Synonym Trifluoromethyl(1,10-phenanthroline)copper(I); Trifluoromethylator|r
    Health Hazard 1 Warning
    Chemical Name or Material (1,10-Phenanthroline)(trifluoromethyl)copper(I)
    Assay 95%

    GHS H Statement
    H302-H312-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P301+P312-P312-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    2° to 8°C

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit