Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (1,10-Phenanthroline)(trifluoromethyl)copper(I), 95%

Manufacturer:  Alfa Aesar™ 4706803

Quantity 1g

 View more versions of this product

Catalog No. AA4706803

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

1300746-79-5
312.76
Trifluoromethyl(1,10-phenanthroline)copper(I); Trifluoromethylator|r
(C12H8N2)CuCF3
MFCD19690901
(1,10-Phenanthroline)(trifluoromethyl)copper(I)

Specifications

1300746-79-5
Orange
Solid
312.76
Trifluoromethyl(1,10-phenanthroline)copper(I); Trifluoromethylator|r
95%
1g
Air & Moisture Sensitive
(C12H8N2)CuCF3
MFCD19690901
(1,10-Phenanthroline)(trifluoromethyl)copper(I)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H312-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P312-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit