Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,12-Dodecanedicarboxylic Acid 98%, Acros Organics

Click to view available options
Quantity:
5g
821-38-5
C14H26O4
258.358
HQHCYKULIHKCEB-UHFFFAOYSA-N
1,12-dodecanedicarboxylic acid,tetradecane-1,14-dioic acid,1,14-tetradecanedioic acid,unii-uec3ac47h1,dodecamethylenedicarboxylic acid,tetradecanedicarboxylic acid,uec3ac47h1,1,12-dodecanedicarboxylicacid,tetradecanedicarboxylate,pubchem18099
13185
CHEBI:76308
tetradecanedioic acid
C(CCCCCCC(=O)O)CCCCCC(=O)O
821-38-5
C14H26O4
258.358
HQHCYKULIHKCEB-UHFFFAOYSA-N
1,12-dodecanedicarboxylic acid,tetradecane-1,14-dioic acid,1,14-tetradecanedioic acid,unii-uec3ac47h1,dodecamethylenedicarboxylic acid,tetradecanedicarboxylic acid,uec3ac47h1,1,12-dodecanedicarboxylicacid,tetradecanedicarboxylate,pubchem18099
13185
CHEBI:76308
tetradecanedioic acid
C(CCCCCCC(=O)O)CCCCCC(=O)O
Color White
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HO2C(CH2)12CO2H
Beilstein 02, 732
Formula Weight 258.35
Physical Form Fine Powder
Percent Purity ≥97.5%
Chemical Name or Material 1, 12-Dodecanedicarboxylic acid, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.