Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,12-Dodecanediol, 99%

Click to view available options
Quantity:
25g
100g
5675-51-4
C12H26O2
202.338
MFCD00004755
GHLKSLMMWAKNBM-UHFFFAOYSA-N
1,12-dodecanediol,dodecamethylene glycol,chembl17440,1,12-dihydroxy dodecane,1,12-dodecandiol,1,12-dodecane diol,acmc-1ambo,1,12-dihydroxydodecane,ksc270c5h
79758
dodecane-1,12-diol
C(CCCCCCO)CCCCCO
5675-51-4
C12H26O2
202.338
MFCD00004755
GHLKSLMMWAKNBM-UHFFFAOYSA-N
1,12-dodecanediol,dodecamethylene glycol,chembl17440,1,12-dihydroxy dodecane,1,12-dodecandiol,1,12-dodecane diol,acmc-1ambo,1,12-dihydroxydodecane,ksc270c5h
79758
dodecane-1,12-diol
C(CCCCCCO)CCCCCO
Applications
1,12-Dodecanediol is used in the polyester polyols for polyurethanes, coatings and inks, adhesives, lastomers, polyesters and co polyesters, polymer cross linkers, pharmaceutical intermediates, fragrances.

Solubility
Soluble in alcohol and warm ether. Insoluble in water and petroleum ether.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Quantity 100g
Melting Point 81°C to 84°C
Boiling Point 188°C to 190°C (12mmHg)
Beilstein 1742760
Formula Weight 202.34
Percent Purity 99%
Chemical Name or Material 1,12-Dodecanediol