Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,2,5,5-Pentamethyl-4-ethyl-3-imidazoline-3-oxide 99%, ACROS Organics™

Click to view available options
Quantity:
250mg
75491-38-2
C10H20N2O
184.283
MFCD00078166
HBZASRFMSFYBQC-UHFFFAOYSA-N
1,2,2,5,5-pentamethyl-4-ethyl-3-imidazoline-3-oxide,5-ethyl-2,2,3,4,4-pentamethylimidazol-1-ium-1-olate,1h-imidazole,4-ethyl-2,5-dihydro-1,2,2,5,5-pentamethyl-, 3-oxide,4-ethyl-1,2,2,5,5-pentamethyl-3-oxo-2,5-dihydro-1h-3lambda~5~-imidazole
2723750
5-ethyl-2,2,3,4,4-pentamethyl-1-oxidoimidazol-1-ium
CCC1=[N+](C(N(C1(C)C)C)(C)C)[O-]
75491-38-2
C10H20N2O
184.283
MFCD00078166
HBZASRFMSFYBQC-UHFFFAOYSA-N
1,2,2,5,5-pentamethyl-4-ethyl-3-imidazoline-3-oxide,5-ethyl-2,2,3,4,4-pentamethylimidazol-1-ium-1-olate,1h-imidazole,4-ethyl-2,5-dihydro-1,2,2,5,5-pentamethyl-, 3-oxide,4-ethyl-1,2,2,5,5-pentamethyl-3-oxo-2,5-dihydro-1h-3lambda~5~-imidazole
2723750
5-ethyl-2,2,3,4,4-pentamethyl-1-oxidoimidazol-1-ium
CCC1=[N+](C(N(C1(C)C)C)(C)C)[O-]
Infrared Spectrum Authentic
Quantity 250mg
Color Colorless
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 184.28
Physical Form Crystals
Percent Purity ≥98.5%
Chemical Name or Material 1,2,2,5,5-Pentamethyl-4-ethyl-3-imidazoline-3-oxide, 99%