Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrafluorobenzene 97%, Acros Organics

Click to view available options
Quantity:
2.5g
551-62-2
C6H2F4
150.076
MFCD00000285
SOZFIIXUNAKEJP-UHFFFAOYSA-N
tetrafluorobenzene,1,2,3,4-tetrafluoro-benzene,unii-66365s2rfq,benzene, 1,2,3,4-tetrafluoro,2,3,4,5-tetrafluorobenzene,benzene, tetrafluoro,pubchem1061,acmc-1ao1z,1,3,4-tetrafluorobenzene
11084
1,2,3,4-tetrafluorobenzene
C1=CC(=C(C(=C1F)F)F)F
551-62-2
C6H2F4
150.076
MFCD00000285
SOZFIIXUNAKEJP-UHFFFAOYSA-N
tetrafluorobenzene,1,2,3,4-tetrafluoro-benzene,unii-66365s2rfq,benzene, 1,2,3,4-tetrafluoro,2,3,4,5-tetrafluorobenzene,benzene, tetrafluoro,pubchem1061,acmc-1ao1z,1,3,4-tetrafluorobenzene
11084
1,2,3,4-tetrafluorobenzene
C1=CC(=C(C(=C1F)F)F)F
Quantity 2.5g
Refractive Index 1.4070 to 1.4090 (20°C, 589nm)
Color Colorless
Density 1.42g/mL
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H2F4
Formula Weight 150.08
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 1,2,3,4-Tetrafluorobenzene, 97%
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger