Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,2,3,4-Tetrafluorobenzene, 99+%

Click to view available options
Quantity:
5g
25g
100g
551-62-2
C6H2F4
150.076
MFCD00000285
SOZFIIXUNAKEJP-UHFFFAOYSA-N
tetrafluorobenzene,1,2,3,4-tetrafluoro-benzene,unii-66365s2rfq,benzene, 1,2,3,4-tetrafluoro,2,3,4,5-tetrafluorobenzene,benzene, tetrafluoro,pubchem1061,acmc-1ao1z,1,3,4-tetrafluorobenzene
11084
1,2,3,4-tetrafluorobenzene
C1=CC(=C(C(=C1F)F)F)F
551-62-2
C6H2F4
150.076
MFCD00000285
SOZFIIXUNAKEJP-UHFFFAOYSA-N
tetrafluorobenzene,1,2,3,4-tetrafluoro-benzene,unii-66365s2rfq,benzene, 1,2,3,4-tetrafluoro,2,3,4,5-tetrafluorobenzene,benzene, tetrafluoro,pubchem1061,acmc-1ao1z,1,3,4-tetrafluorobenzene
11084
1,2,3,4-tetrafluorobenzene
C1=CC(=C(C(=C1F)F)F)F
Boiling Point 94°C to 96°C
Melting Point -42°C
Quantity 25g
Flash Point 20°C (69°F)
UN Number UN1993
Beilstein 1910024
Formula Weight 150.07
Percent Purity ≥99%
Chemical Name or Material 1,2,3,4-Tetrafluorobenzene

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.