Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,2,3,4-Tetrahydrocarbazole, 99%

Click to view available options
Quantity:
25g
942-01-8
C12H13N
171.243
MFCD00004959
XKLNOVWDVMWTOB-UHFFFAOYSA-N
1,2,3,4-tetrahydrocarbazole,2,3-tetramethyleneindole,2,3-tetramethylene-1h-indole,5,6,7,8-tetrahydrocarbazole,tetrahydrocarbazole,1h-carbazole, 2,3,4,9-tetrahydro,carbazole, 1,2,3,4-tetrahydro,1h-indole, 2,3-1,4-butanediyl,carbazole, 5,6,7,8-tetrahydro,unii-8zlk0tsx93
13664
2,3,4,9-tetrahydro-1H-carbazole
C1CCC2=C(C1)C3=CC=CC=C3N2
942-01-8
C12H13N
171.243
MFCD00004959
XKLNOVWDVMWTOB-UHFFFAOYSA-N
1,2,3,4-tetrahydrocarbazole,2,3-tetramethyleneindole,2,3-tetramethylene-1h-indole,5,6,7,8-tetrahydrocarbazole,tetrahydrocarbazole,1h-carbazole, 2,3,4,9-tetrahydro,carbazole, 1,2,3,4-tetrahydro,1h-indole, 2,3-1,4-butanediyl,carbazole, 5,6,7,8-tetrahydro,unii-8zlk0tsx93
13664
2,3,4,9-tetrahydro-1H-carbazole
C1CCC2=C(C1)C3=CC=CC=C3N2
Applications
It finds its application as a pharmaceutical intermediate and in the synthesis of aspidospermidine alkaloids.

Solubility
Insoluble in water. Solubility in methanol (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Boiling Point 325°C to 330°C
Quantity 25g
Melting Point 118°C to 120°C
Beilstein 133771
Formula Weight 171.24
Percent Purity 99%
Chemical Name or Material 1,2,3,4-Tetrahydrocarbazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : FE6300000