Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydroisoquinoline hydrochloride, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
14099-81-1
C9H12ClN
169.652
MFCD00012740
MGFREDWKELGWML-UHFFFAOYSA-N
1,2,3,4-tetrahydroisoquinoline hydrochloride,tetrahydroisoquinoline hydrochloride,1,2,3,4-tetrahydro-isoquinoline hcl,1,2,3,4-tetrahydroisoquinoline, chloride,acmc-209clj,1,2,3,4-tetrahydroisoquinoline, hydrochloride,isoquinoline,1,2,3,4-tetrahydro-, hydrochloride 1:1
2734717
1,2,3,4-tetrahydroisoquinoline;hydrochloride
C1CNCC2=CC=CC=C21.Cl
14099-81-1
C9H12ClN
169.652
MFCD00012740
MGFREDWKELGWML-UHFFFAOYSA-N
1,2,3,4-tetrahydroisoquinoline hydrochloride,tetrahydroisoquinoline hydrochloride,1,2,3,4-tetrahydro-isoquinoline hcl,1,2,3,4-tetrahydroisoquinoline, chloride,acmc-209clj,1,2,3,4-tetrahydroisoquinoline, hydrochloride,isoquinoline,1,2,3,4-tetrahydro-, hydrochloride 1:1
2734717
1,2,3,4-tetrahydroisoquinoline;hydrochloride
C1CNCC2=CC=CC=C21.Cl
Quantity 5g
Color White
Melting Point 198°C to 202°C
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 169.65
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 1,2,3,4-Tetrahydroisoquinoline hydrochloride
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.