Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetrahydroquinoline, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
2.5kg
Packaging:
Glass bottle
635-46-1
C9H11N
133.19
LBUJPTNKIBCYBY-UHFFFAOYSA-N
tetrahydroquinoline,1,2,3,4-tetrahydro-quinoline,kusol,quinoline, 1,2,3,4-tetrahydro,unii-ccr50n1z9g,1,2,3,4-tetrahydrochinoline,1,2,3,4-tetrahydro quinoline,benzopiperidine,ccr50n1z9g,1,2,3,4,-tetrahydroquinoline
69460
CHEBI:213323
1,2,3,4-tetrahydroquinoline
C1CC2=CC=CC=C2NC1
635-46-1
C9H11N
133.19
LBUJPTNKIBCYBY-UHFFFAOYSA-N
tetrahydroquinoline,1,2,3,4-tetrahydro-quinoline,kusol,quinoline, 1,2,3,4-tetrahydro,unii-ccr50n1z9g,1,2,3,4-tetrahydrochinoline,1,2,3,4-tetrahydro quinoline,benzopiperidine,ccr50n1z9g,1,2,3,4,-tetrahydroquinoline
69460
CHEBI:213323
1,2,3,4-tetrahydroquinoline
C1CC2=CC=CC=C2NC1
Viscosity 12.5 mPa.s (20°C)
Density 1.0600g/mL
Infrared Spectrum Authentic
Flash Point 100°C
Specific Gravity 1.06
Color Yellow
Packaging Glass bottle
Melting Point 15.0°C to 17.0°C
Boiling Point 249.0°C
Quantity 25g
Refractive Index 1.5930 to 1.5950
Assay Percent Range 98%
Beilstein 20,262
Solubility Information Solubility in water: <1g/L (20°C). Other solubilities: soluble in ethanol
Formula Weight 133.19
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1,2,3,4-Tetrahydroquinoline
Show More Show Less

Danger

Recommended Storage : Product may darken during storage