Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Tetramethyl-1,3-cyclopentadiene, 85%, mixture of isomers, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
Quantity 5g
Boiling Point 142.0°C
Flash Point 42°C
Infrared Spectrum Authentic
Color Yellow
Density 0.8080g/mL
Specific Gravity 0.808
Packaging Glass bottle
Refractive Index 1.4700 to 1.4760
CAS 4249-10-9
Assay Percent Range 80% min. (GC)
Molecular Formula C9H14
Synonym 1,2,3,4-tetramethylcyclopentadiene,tetramethylcyclopentadiene,1,2,3,4-tetramethyl-1,3-cyclopentadiene,tetramethyl cyclopentadiene,tetramethylcyclopentadiene, mixed isomers,pubchem15072,acmc-1an6q,1,3-cyclopentadiene, tetramethyl,2,3,4,5-tetramethylcyclopenta-2,4-diene
InChI Key VNPQQEYMXYCAEZ-UHFFFAOYSA-N
SMILES CC1=C(C(=C(C1)C)C)C
IUPAC Name 1,2,3,4-tetramethylcyclopenta-1,3-diene
Molecular Weight (g/mol) 122.21
PubChem CID 138163
Formula Weight 122.21
Physical Form Liquid
Percent Purity 85%
Chemical Name or Material 1,2,3,4-Tetramethyl-1,3-cyclopentadiene
Show More Show Less

GHS Signal Word: Warning