Alfa Aesar™ 1-(2,3,5,6-Tetramethylphenylsulfonyl)-L-proline, 96%

Manufacturer: Alfa Aesar™ H3381903

Quantity 1g

 View more versions of this product

Catalog No. AAH3381903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1009282-06-7
    Molecular Formula C15H21NO4S
    Formula Weight 311.4
    MDL Number MFCD06409475
    Chemical Name or Material 1-(2,3,5,6-Tetramethylphenylsulfonyl)-L-proline

    Specifications

    Formula Weight 311.4
    Molecular Formula C15H21NO4S
    MDL Number MFCD06409475
    CAS 1009282-06-7
    Melting Point 121° to 124°C
    Quantity 1g
    Optical Rotation −47° (c=1 in DMF)
    Chemical Name or Material 1-(2,3,5,6-Tetramethylphenylsulfonyl)-L-proline
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit