Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,2,3-Benzenetricarboxylic acid hydrate, 99%

Click to view available options
Quantity:
5g
10g
25g
732304-21-1
C9H8O7
228.156
MFCD00149097
DYDNZUYNRZENMU-UHFFFAOYSA-N
1,2,3-benzenetricarboxylic acid hydrate,hemimellitic acid hydrate,benzene-1,2,3-tricarboxylic acid, oxamethane,acmc-1ajgh
16219028
benzene-1,2,3-tricarboxylic acid;hydrate
C1=CC(=C(C(=C1)C(=O)O)C(=O)O)C(=O)O.O
732304-21-1
C9H8O7
228.156
MFCD00149097
DYDNZUYNRZENMU-UHFFFAOYSA-N
1,2,3-benzenetricarboxylic acid hydrate,hemimellitic acid hydrate,benzene-1,2,3-tricarboxylic acid, oxamethane,acmc-1ajgh
16219028
benzene-1,2,3-tricarboxylic acid;hydrate
C1=CC(=C(C(=C1)C(=O)O)C(=O)O)C(=O)O.O
Applications
It is used in the synthesis and characterization and thermal behavior of hemimellitic acid complexes with lanthanides(III).

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Melting Point ~191°C (decomposition)
Quantity 5g
Beilstein 2214816
Solubility Information Slightly soluble in water.
Formula Weight 210.14 (Anhydrous)
Percent Purity 99%
Chemical Name or Material 1,2,3-Benzenetricarboxylic acid hydrate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-317-0

TSCA : No

Recommended Storage : Ambient temperatures