Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2,3-Dimethylphenyl)piperazine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
1013-22-5
C12H18N2
190.29
LIKXJDINUMWKQA-UHFFFAOYSA-N
1-2,3-dimethylphenyl piperazine,1-2,3-xylyl piperazine,piperazine, 1-2,3-dimethylphenyl,1-2,3-dimethylphenyl-piperazine,piperazine,1-2,3-dimethylphenyl,2,3-dimethylphenyl piperazine,1-2,3-xylyl piperazine hydrochloride,pubchem8587,acmc-1bppt,4-2,3-dimethylphenyl-piperazine
70541
1-(2,3-dimethylphenyl)piperazine
CC1=C(C(=CC=C1)N2CCNCC2)C
1013-22-5
C12H18N2
190.29
LIKXJDINUMWKQA-UHFFFAOYSA-N
1-2,3-dimethylphenyl piperazine,1-2,3-xylyl piperazine,piperazine, 1-2,3-dimethylphenyl,1-2,3-dimethylphenyl-piperazine,piperazine,1-2,3-dimethylphenyl,2,3-dimethylphenyl piperazine,1-2,3-xylyl piperazine hydrochloride,pubchem8587,acmc-1bppt,4-2,3-dimethylphenyl-piperazine
70541
1-(2,3-dimethylphenyl)piperazine
CC1=C(C(=CC=C1)N2CCNCC2)C
Refractive Index 1.5566 to 1.5606
Packaging Glass bottle
Color Yellow
Quantity 25g
Infrared Spectrum Authentic
Density 1.0309g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 190.29
Physical Form Viscous Liquid
Percent Purity 99%
Chemical Name or Material 1-(2, 3-Dimethylphenyl)piperazine
Show More Show Less

GHS H Statement
Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning