Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,2,3-Trichloro-5-nitrobenzene, 97%

Click to view available options
Quantity:
5g
25g
20098-48-0
C6H2Cl3NO2
226.437
MFCD00061096
HHLCSFGOTLUREE-UHFFFAOYSA-N
3,4,5-trichloronitrobenzene,benzene, 1,2,3-trichloro-5-nitro,5-nitro-1,2,3-trichlorobenzene,unii-ah0cu8672k,1,2,3-trichloro-5-nitro-benzene,pubchem3717,acmc-1cmcw,3,5-trichloronitrobenzene,dsstox_cid_6205,dsstox_rid_78057
88365
1,2,3-trichloro-5-nitrobenzene
C1=C(C=C(C(=C1Cl)Cl)Cl)[N+](=O)[O-]
20098-48-0
C6H2Cl3NO2
226.437
MFCD00061096
HHLCSFGOTLUREE-UHFFFAOYSA-N
3,4,5-trichloronitrobenzene,benzene, 1,2,3-trichloro-5-nitro,5-nitro-1,2,3-trichlorobenzene,unii-ah0cu8672k,1,2,3-trichloro-5-nitro-benzene,pubchem3717,acmc-1cmcw,3,5-trichloronitrobenzene,dsstox_cid_6205,dsstox_rid_78057
88365
1,2,3-trichloro-5-nitrobenzene
C1=C(C=C(C(=C1Cl)Cl)Cl)[N+](=O)[O-]
Quantity 25g
Melting Point 68°C to 71°C
UN Number UN2811
Beilstein 2505480
Formula Weight 226.45
Percent Purity 97%
Chemical Name or Material 1,2,3-Trichloro-5-nitrobenzene

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.