Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,2,3-Trichlorobenzene, 99%

Click to view available options
Quantity:
100g
500g
87-61-6
C6H3Cl3
181.44
MFCD00000537
RELMFMZEBKVZJC-UHFFFAOYSA-N
trichlorobenzene,vic-trichlorobenzene,1,2,6-trichlorobenzene,benzene, 1,2,3-trichloro,benzene, trichloro,unii-nur9777ik4,ccris 5944,dsstox_cid_6193,dsstox_rid_78052,dsstox_gsid_26193
6895
CHEBI:35289
1,2,3-trichlorobenzene
C1=CC(=C(C(=C1)Cl)Cl)Cl
87-61-6
C6H3Cl3
181.44
MFCD00000537
RELMFMZEBKVZJC-UHFFFAOYSA-N
trichlorobenzene,vic-trichlorobenzene,1,2,6-trichlorobenzene,benzene, 1,2,3-trichloro,benzene, trichloro,unii-nur9777ik4,ccris 5944,dsstox_cid_6193,dsstox_rid_78052,dsstox_gsid_26193
6895
CHEBI:35289
1,2,3-trichlorobenzene
C1=CC(=C(C(=C1)Cl)Cl)Cl
Applications
Detoxification by catalytic hydrotreatment of 1,2,3-Trichlorobenzene is used for the disposal of hazardous organic waste liquids. As constituent of trichlorobenzene mixt used for termite control. As transformer fluid, dye carrier & solvent. Solvent for high melting products, Coolant in electrical installations and glass tempering. In polyester dyeing, lubricants, Heat transfer medium. As chemical intermediate for 2,3-dichlorophenol.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Melting Point 53°C to 55°C
Quantity 100g
Boiling Point 218°C to 219°C
Flash Point 126°C (258°F)
UN Number UN3077
Beilstein 956882
Merck Index 14,9630
Formula Weight 181.45
Percent Purity 99%
Chemical Name or Material 1,2,3-Trichlorobenzene

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : DC2095000

TSCA : TSCA