Alfa Aesar™ 1,2,3-Trichlorobenzene, 99%

Manufacturer: Alfa Aesar™ B2215436

Quantity 500g

 View more versions of this product

Catalog No. AAB2215436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 87-61-6
    Molecular Formula C6H3Cl3
    Formula Weight 181.45
    MDL Number MFCD00000537
    UN Number UN3077
    Beilstein 956882
    Merck Index 14,9630
    Chemical Name or Material 1,2,3-Trichlorobenzene

    Specifications

    Formula Weight 181.45
    Melting Point 53° to 55°C
    Quantity 500g
    TSCA TSCA
    Molecular Formula C6H3Cl3
    MDL Number MFCD00000537
    UN Number UN3077
    Merck Index 14,9630
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301
    Toxic if swallowed.
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 218° to 219°C
    Flash Point 126°C (258°F)
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    RTECS Number DC2095000
    CAS 87-61-6
    Beilstein 956882
    Chemical Name or Material 1,2,3-Trichlorobenzene
    Assay 99%

    Description

    Applications
    Detoxification by catalytic hydrotreatment of 1,2,3-Trichlorobenzene is used for the disposal of hazardous organic waste liquids. As constituent of trichlorobenzene mixt used for termite control. As transformer fluid, dye carrier & solvent. Solvent for high melting products, Coolant in electrical installations and glass tempering. In polyester dyeing, lubricants, Heat transfer medium. As chemical intermediate for 2,3-dichlorophenol.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H301
    Toxic if swallowed.

    GHS P Statement
    P264-P270-P301+P310-P321-P405-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Specific treatment (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit