Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Trichlorobenzene 99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
87-61-6
C6H3Cl3
181.44
MFCD00000537
RELMFMZEBKVZJC-UHFFFAOYSA-N
trichlorobenzene,vic-trichlorobenzene,1,2,6-trichlorobenzene,benzene, 1,2,3-trichloro,benzene, trichloro,unii-nur9777ik4,ccris 5944,dsstox_cid_6193,dsstox_rid_78052,dsstox_gsid_26193
6895
CHEBI:35289
1,2,3-trichlorobenzene
C1=CC(=C(C(=C1)Cl)Cl)Cl
87-61-6
C6H3Cl3
181.44
MFCD00000537
RELMFMZEBKVZJC-UHFFFAOYSA-N
trichlorobenzene,vic-trichlorobenzene,1,2,6-trichlorobenzene,benzene, 1,2,3-trichloro,benzene, trichloro,unii-nur9777ik4,ccris 5944,dsstox_cid_6193,dsstox_rid_78052,dsstox_gsid_26193
6895
CHEBI:35289
1,2,3-trichlorobenzene
C1=CC(=C(C(=C1)Cl)Cl)Cl
Melting Point 50°C to 54°C
Packaging Glass bottle
Color White
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 05, 203
Merck Index 15, 9795
Formula Weight 181.45
Physical Form Crystalline Solid
Percent Purity ≥98.5%
Chemical Name or Material 1, 2, 3-Trichlorobenzene, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Very toxic to aquatic life with long lasting effects.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-757-1

RTECSNumber : DC2095000

TSCA : TSCA