Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Trichloropropane, 98+%, ACROS Organics™

Click to view available options
Quantity:
5mL
250mL
1L
Packaging:
Glass bottle
96-18-4
C3H5Cl3
147.423
MFCD00000946
CFXQEHVMCRXUSD-UHFFFAOYSA-N
trichlorohydrin,allyl trichloride,propane, 1,2,3-trichloro,glycerol trichlorohydrin,glyceryl trichlorohydrin,unii-3mj7qck0z0,ccris 5874,3mj7qck0z0,dsstox_cid_1390,dsstox_rid_76132
7285
CHEBI:34036
1,2,3-trichloropropane
C(C(CCl)Cl)Cl
96-18-4
C3H5Cl3
147.423
MFCD00000946
CFXQEHVMCRXUSD-UHFFFAOYSA-N
trichlorohydrin,allyl trichloride,propane, 1,2,3-trichloro,glycerol trichlorohydrin,glyceryl trichlorohydrin,unii-3mj7qck0z0,ccris 5874,3mj7qck0z0,dsstox_cid_1390,dsstox_rid_76132
7285
CHEBI:34036
1,2,3-trichloropropane
C(C(CCl)Cl)Cl
Quantity 5mL
Refractive Index 1.4830 to 1.4850 (20°C, 589nm)
Color Colorless to Yellow
Packaging Glass bottle
Density 1.3860g/mL
Infrared Spectrum Authentic
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula ClCH2CH(Cl)CH2Cl
Beilstein 01, 106
Formula Weight 147.43
Physical Form Liquid
Percent Purity ≥98.0%
Chemical Name or Material 1, 2, 3-Trichloropropane
Show More Show Less

GHS H Statement:
May cause cancer.
May damage fertility.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger