Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Trichloropropane 98.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
96-18-4
C3H5Cl3
147.423
MFCD00000946
CFXQEHVMCRXUSD-UHFFFAOYSA-N
trichlorohydrin,allyl trichloride,propane, 1,2,3-trichloro,glycerol trichlorohydrin,glyceryl trichlorohydrin,unii-3mj7qck0z0,ccris 5874,3mj7qck0z0,dsstox_cid_1390,dsstox_rid_76132
7285
CHEBI:34036
1,2,3-trichloropropane
C(C(CCl)Cl)Cl
96-18-4
C3H5Cl3
147.423
MFCD00000946
CFXQEHVMCRXUSD-UHFFFAOYSA-N
trichlorohydrin,allyl trichloride,propane, 1,2,3-trichloro,glycerol trichlorohydrin,glyceryl trichlorohydrin,unii-3mj7qck0z0,ccris 5874,3mj7qck0z0,dsstox_cid_1390,dsstox_rid_76132
7285
CHEBI:34036
1,2,3-trichloropropane
C(C(CCl)Cl)Cl
Boiling Point 156°C
Color Yellow
Quantity 500g
UN Number 2810
Formula Weight 147.42
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material 1,2,3-Trichloropropane

EINECSNumber : (2)-0083

RTECSNumber : TZ9275000

TSCA : Yes