Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-1H-Triazole, 99.5%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass Bottle
Plastic bottle
288-88-0
C2H3N3
69.06
MFCD00005228
NSPMIYGKQJPBQR-UHFFFAOYSA-N
1,2,4-triazole,4h-1,2,4-triazole,pyrrodiazole,s-triazole,1,2,4-1h-triazole,1h-1,2,4-triazol,unii-10ms0y1rdi,1,2,4 triazole,1,2,4-triazol,4h-1,2,4-triazole van
9257
CHEBI:35550
1H-1,2,4-triazole
C1=NC=NN1
288-88-0
C2H3N3
69.06
MFCD00005228
NSPMIYGKQJPBQR-UHFFFAOYSA-N
1,2,4-triazole,4h-1,2,4-triazole,pyrrodiazole,s-triazole,1,2,4-1h-triazole,1h-1,2,4-triazol,unii-10ms0y1rdi,1,2,4 triazole,1,2,4-triazol,4h-1,2,4-triazole van
9257
CHEBI:35550
1H-1,2,4-triazole
C1=NC=NN1
Flash Point 140°C
Boiling Point 260°C
Quantity 100g
Infrared Spectrum Authentic
Packaging Plastic bottle
Water 0.2% max. (K.F.)
Melting Point 119°C to 122°C
Color White
CAS Min % 99.4
CAS Max % 100.0
Assay Percent Range 99.5%
Beilstein 26, 13
Fieser 01,1188; 02,423
Merck Index 15, 9766
Solubility Information Solubility in water: 1250g/L (20°C). Other solubilities: soluble in chloroform,xylene,ethylacetate and,alcohol
Formula Weight 69.06
Physical Form Crystalline Powder and Flakes
Percent Purity ≥99.4%
Chemical Name or Material 1, 2, 4-1H-Triazole
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Suspected of damaging the unborn child.

GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.