Alfa Aesar™ 1,2,4,5-Tetrakis(isopropylthio)benzene, 97%

Manufacturer: Alfa Aesar™ B2526106

Quantity 5g

 View more versions of this product

Catalog No. AAB2526106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 74542-69-1
    Molecular Formula C18H30S4
    Formula Weight 374.68
    MDL Number MFCD04039784
    Synonym 1,2,4,5-Tetra(isopropylthio)benzene
    Chemical Name or Material 1,2,4,5-Tetrakis(isopropylthio)benzene

    Specifications

    Formula Weight 374.68
    MDL Number MFCD04039784
    Synonym 1,2,4,5-Tetra(isopropylthio)benzene
    Quantity 5g
    Molecular Formula C18H30S4
    CAS 74542-69-1
    Melting Point 76° to 78°C
    Chemical Name or Material 1,2,4,5-Tetrakis(isopropylthio)benzene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit