Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4,5-Tetramethylbenzene, 97+%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
95-93-2
C10H14
134.22
SQNZJJAZBFDUTD-UHFFFAOYSA-N
durene,durol,benzene, 1,2,4,5-tetramethyl,unii-181426cfyb,p-xylene, 2,5-dimethyl,dsstox_cid_9124,dsstox_rid_78675,dsstox_gsid_29124,duren
7269
CHEBI:38978
1,2,4,5-tetramethylbenzene
CC1=CC(=C(C=C1C)C)C
95-93-2
C10H14
134.22
SQNZJJAZBFDUTD-UHFFFAOYSA-N
durene,durol,benzene, 1,2,4,5-tetramethyl,unii-181426cfyb,p-xylene, 2,5-dimethyl,dsstox_cid_9124,dsstox_rid_78675,dsstox_gsid_29124,duren
7269
CHEBI:38978
1,2,4,5-tetramethylbenzene
CC1=CC(=C(C=C1C)C)C
Flash Point 73°C
Boiling Point 196.0°C to 197.0°C
Melting Point 78.0°C to 82.0°C
Infrared Spectrum Authentic
Quantity 100g
Assay Percent Range 97% min. (GC)
Linear Formula C6H2(CH3)4
Beilstein 05,431
Merck Index 15,3515
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 134.22
Percent Purity 97+%
Chemical Name or Material 1,2,4,5-Tetramethylbenzene
Show More Show Less

GHS H Statement
Flammable solid.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environment.

GHS Signal Word: Danger

EINECSNumber : 202-465-7