Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4,5-Tetramethylbenzene, 97+%, Acros Organics™

Click to view available options
Quantity:
5g
100g
500g
95-93-2
C10H14
134.222
SQNZJJAZBFDUTD-UHFFFAOYSA-N
durene,durol,benzene, 1,2,4,5-tetramethyl,unii-181426cfyb,p-xylene, 2,5-dimethyl,dsstox_cid_9124,dsstox_rid_78675,dsstox_gsid_29124,duren
7269
CHEBI:38978
1,2,4,5-tetramethylbenzene
CC1=CC(=C(C=C1C)C)C
95-93-2
C10H14
134.222
SQNZJJAZBFDUTD-UHFFFAOYSA-N
durene,durol,benzene, 1,2,4,5-tetramethyl,unii-181426cfyb,p-xylene, 2,5-dimethyl,dsstox_cid_9124,dsstox_rid_78675,dsstox_gsid_29124,duren
7269
CHEBI:38978
1,2,4,5-tetramethylbenzene
CC1=CC(=C(C=C1C)C)C
Melting Point 78.0°C to 82.0°C
Infrared Spectrum Authentic
Flash Point 73°C
Quantity 500g
Boiling Point 196.0°C to 197.0°C
Linear Formula C6H2(CH3)4
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 134.22
Percent Purity ≥97%
Chemical Name or Material 1,2,4,5-Tetramethylbenzene

GHS H Statement Flammable solid. Very toxic to aquatic life with long lasting effects.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid release to the environment.

Danger