Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Benzenetricarboxylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
250g
Packaging:
Glass Bottle
Plastic Bottle
528-44-9
C9H6O6
210.141
MFCD00002470
ARCGXLSVLAOJQL-UHFFFAOYSA-N
1,2,4-benzenetricarboxylic acid,trimellitic acid,1,2,4-tricarboxybenzene,4-carboxyphthalic acid,1,3,4-benzenetricarboxylic acid,1,4,5-benzenetricarboxylic acid,1,2,4-benzenetricarboxylicacid,unii-7nvy29mq5f,7nvy29mq5f,dsstox_cid_1487
10708
CHEBI:166055
benzene-1,2,4-tricarboxylic acid
C1=CC(=C(C=C1C(=O)O)C(=O)O)C(=O)O
528-44-9
C9H6O6
210.141
MFCD00002470
ARCGXLSVLAOJQL-UHFFFAOYSA-N
1,2,4-benzenetricarboxylic acid,trimellitic acid,1,2,4-tricarboxybenzene,4-carboxyphthalic acid,1,3,4-benzenetricarboxylic acid,1,4,5-benzenetricarboxylic acid,1,2,4-benzenetricarboxylicacid,unii-7nvy29mq5f,7nvy29mq5f,dsstox_cid_1487
10708
CHEBI:166055
benzene-1,2,4-tricarboxylic acid
C1=CC(=C(C=C1C(=O)O)C(=O)O)C(=O)O
Packaging Plastic Bottle
Color White
Infrared Spectrum Authentic
Quantity 50g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H3(COH)3
Beilstein 09, 977
Merck Index 15, 9871
Formula Weight 210.14
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 1, 2, 4-Benzenetricarboxylic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.