Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2,4-Dichlorophenyl)-1-cyclopropyl Cyanide 98%, ACROS Organics™

Click to view available options
Quantity:
25g
71463-55-3
C10H7Cl2N
212.08
MFCD00044818
VDKKBWVLLSVWKI-UHFFFAOYSA-N
1-2,4-dichlorophenyl cyclopropanecarbonitrile,1-2,4-dichlorophenyl cyclopropane-1-carbonitrile,1-2,4-dichlorophenyl-1-cyclopropyl cyanide,1-2,4-dichlorophenyl-1-cyanocyclopropane,1-2,4-dichloro-phenyl-cyclopropanecarbonitrile,cyclopropanecarbonitrile, 1-2,4-dichlorophenyl
1268258
1-(2,4-dichlorophenyl)cyclopropane-1-carbonitrile
C1CC1(C#N)C2=C(C=C(C=C2)Cl)Cl
71463-55-3
C10H7Cl2N
212.08
MFCD00044818
VDKKBWVLLSVWKI-UHFFFAOYSA-N
1-2,4-dichlorophenyl cyclopropanecarbonitrile,1-2,4-dichlorophenyl cyclopropane-1-carbonitrile,1-2,4-dichlorophenyl-1-cyclopropyl cyanide,1-2,4-dichlorophenyl-1-cyanocyclopropane,1-2,4-dichloro-phenyl-cyclopropanecarbonitrile,cyclopropanecarbonitrile, 1-2,4-dichlorophenyl
1268258
1-(2,4-dichlorophenyl)cyclopropane-1-carbonitrile
C1CC1(C#N)C2=C(C=C(C=C2)Cl)Cl
Melting Point 81°C to 85°C
Packaging Glass Bottle
Quantity 25g
Assay Percent Range 98%
Formula Weight 212.08
Chemical Name or Material 1-(2, 4-Dichlorophenyl)-1-cyclopropyl cyanide, 98%