Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-(2,4-Dimethylphenyl)piperazine, 99%, Thermo Scientific™
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.5530 to 1.555 |
Quantity | 5 g |
Formula Weight | 190.29 |
Percent Purity | 99% |
Physical Form | Liquid |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Harmful if swallowed. Harmful in contact with skin. Causes severe skin burns and eye damage. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present a |
EINECSNumber | 213-797-7 |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction