Alfa Aesar™ 1,2,4-Triacetoxybenzene, 97%

Manufacturer: Alfa Aesar™ B2420814

Quantity 25g

 View more versions of this product

Catalog No. AAB2420814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 613-03-6
    Molecular Formula C12H12O6
    Formula Weight 252.22
    MDL Number MFCD00008700
    Beilstein 2138876
    Chemical Name or Material 1,2,4-Triacetoxybenzene

    Specifications

    Formula Weight 252.22
    TSCA TSCA
    Molecular Formula C12H12O6
    MDL Number MFCD00008700
    CAS 613-03-6
    Beilstein 2138876
    Melting Point 98° to 100°C
    Quantity 25g
    RTECS Number DC4800000
    Chemical Name or Material 1,2,4-Triacetoxybenzene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit